Anti-COX17 Mouse monoclonal antibody Biotin [clone: 10B2583]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-89W-BIOTEA 0 DKK
USBIC7904-89W-BIOT
Anti-COX17 Mouse monoclonal antibody Biotin [clone: 10B2583]
Type: Primary
Antigen: COX17 (COX17 cytochrome c oxidase copper chaperone)
Clonality: Monoclonal
Clone: 10B2583
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR