Anti-COX17 Mouse Monoclonal Antibody [clone: 4G2]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-89W1EA 4790 DKK
USBIC7904-89W1
Anti-COX17 Mouse Monoclonal Antibody [clone: 4G2]
Antistoffer
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: COX17 (COX17 cytochrome c oxidase copper chaperone)
Clonality: Monoclonal
Clone: 4G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR