Anti-COX6C Mouse monoclonal antibody Biotin [clone: 10B2587]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-89N-BIOTEA 0 DKK
USBIC7904-89N-BIOT
Anti-COX6C Mouse monoclonal antibody Biotin [clone: 10B2587]
Type: Primary
Antigen: COX6C (cytochrome c oxidase subunit VIc)
Clonality: Monoclonal
Clone: 10B2587
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR