Anti-COX5B Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2585]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-89H-APEA 0 DKK
USBIC7904-89H-AP
Anti-COX5B Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2585]
Type: Primary
Antigen: COX5B (cytochrome c oxidase subunit Vb)
Clonality: Monoclonal
Clone: 10B2585
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR