Anti-COX4I2 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-89FEA 3460 DKK
USBIC7904-89F
Anti-COX4I2 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1: 2000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: COX4I2 (cytochrome c oxidase subunit IV isoform 2 (lung))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR