Anti-COX4I2 Mouse Monoclonal Antibody [clone: 12K71]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-89E2EA 4320 DKK
USBIC7904-89E2
Anti-COX4I2 Mouse Monoclonal Antibody [clone: 12K71]
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: COX4I2 (cytochrome c oxidase subunit IV isoform 2 (lung))
Clonality: Monoclonal
Clone: 12K71
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR