Anti-COX 2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-87CEA 2150 DKK
USBIC7904-87C
Anti-COX 2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 2-4?g/ml; Immunochemistry: 1:100

Type: Primary
Antigen: COX 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR