Anti-COX 2 Rabbit Monoclonal Antibody [clone: 5G85]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-87BEA 4510 DKK
USBIC7904-87B
Anti-COX 2 Rabbit Monoclonal Antibody [clone: 5G85]
Antistoffer
Type: Primary
Antigen: COX 2
Clonality: Monoclonal
Clone: 5G85
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR