Anti-Cox 2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-84HEA 5010 DKK
USBIC7904-84H
Anti-Cox 2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:10000-1:50000; Western Blot: 1:5000-1:10000

Type: Primary
Antigen: Cox 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR