Anti-Cyclooxygenase-2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-83MEA 4320 DKK
USBIC7904-83M
Anti-Cyclooxygenase-2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:10000-1:50000; Western Blot: 1:5000-1:10000; Immunochemistry: 1:500

Type: Primary
Antigen: Cyclooxygenase-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR