Anti-COX 2 Mouse Monoclonal Antibody [clone: 6A53]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-83DEA 4040 DKK
USBIC7904-83D
Anti-COX 2 Mouse Monoclonal Antibody [clone: 6A53]
Antistoffer
Type: Primary
Antigen: COX 2
Clonality: Monoclonal
Clone: 6A53
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR