Anti-COX 2 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-80FEA 4930 DKK
USBIC7904-80F
Anti-COX 2 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: COX 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR