Anti-COX 1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-75DEA 3290 DKK
USBIC7904-75D
Anti-COX 1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.1-0.5 ?g/ml; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: COX 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR