Anti-COX 1 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-73CEA 4120 DKK
USBIC7904-73C
Anti-COX 1 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:8000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: COX 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR