Anti-COX 1 Rabbit Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7904-71VEA 4120 DKK
USBIC7904-71V
Anti-COX 1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: COX 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Pig, Sheep