Anti-Corynebacterium diphtheriae toxoid Mouse Monoclonal Antibody [clone: 6F91]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-40EA 5930 DKK
USBIC7904-40
Anti-Corynebacterium diphtheriae toxoid Mouse Monoclonal Antibody [clone: 6F91]
Antistoffer
Type: Primary
Antigen: Corynebacterium diphtheriae toxoid
Clonality: Monoclonal
Clone: 6F91
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR