Anti-Cortistatin-29 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-38LEA 5750 DKK
USBIC7904-38L
Anti-Cortistatin-29 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Cortistatin-29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR