Anti-Cortisol Mouse Monoclonal Antibody [clone: 10B2298]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7904-11CEA 4590 DKK
USBIC7904-11C
Anti-Cortisol Mouse Monoclonal Antibody [clone: 10B2298]
Antistoffer
Type: Primary
Antigen: Cortisol
Clonality: Monoclonal
Clone: 10B2298
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR