Anti-Corticotropin-releasing Factor Receptor 2 Rabbit Polyclonal Antibody (Carboxyfluorescein)
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7903-90D2EA 4910 DKK
USBIC7903-90D2
Anti-Corticotropin-releasing Factor Receptor 2 Rabbit Polyclonal Antibody (Carboxyfluorescein)
Antistoffer
Type: Primary
Antigen: Corticotropin-releasing Factor Receptor 2
Clonality: Polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human