Anti-Corticotropin-releasing Factor Receptor 2 Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7903-90D1EA 4340 DKK
USBIC7903-90D1
Anti-Corticotropin-releasing Factor Receptor 2 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Corticotropin-releasing Factor Receptor 2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR