Anti-CORTICOTROPIN RELEASING FACTOR RECEPTOR 1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7903-89HEA 4320 DKK
USBIC7903-89H
Anti-CORTICOTROPIN RELEASING FACTOR RECEPTOR 1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: CORTICOTROPIN RELEASING FACTOR RECEPTOR 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR