Anti-CORTICOTROPIN RELEASING FACTOR RECEPTOR 1 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7903-89EEA 4540 DKK
USBIC7903-89E
Anti-CORTICOTROPIN RELEASING FACTOR RECEPTOR 1 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:32000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: CORTICOTROPIN RELEASING FACTOR RECEPTOR 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR