Anti-Corticotropin-Releasing Factor Receptor 1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 7H27]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7903-89AEA 4500 DKK
USBIC7903-89A
Anti-Corticotropin-Releasing Factor Receptor 1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 7H27]
Antistoffer
Do not freeze!

Type: Primary
Antigen: Corticotropin-Releasing Factor Receptor 1
Clonality: Monoclonal
Clone: 7H27
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR