Anti-Corticoliberin, Pro Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7903-80ZEA 4500 DKK
USBIC7903-80Z
Anti-Corticoliberin, Pro Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: Corticoliberin, Pro
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR