Anti-Corticotropin Releasing Factor Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7903-80WEA 2660 DKK
USBIC7903-80W
Anti-Corticotropin Releasing Factor Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Corticotropin Releasing Factor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR