Anti-CRHR1 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7903-79CEA 3460 DKK
USBIC7903-79C
Anti-CRHR1 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:32000; Western Blot:

Type: Primary
Antigen: CRHR1 (corticotropin releasing hormone receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR