Anti-CORO1C Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7901-38DEA 3460 DKK
USBIC7901-38D
Anti-CORO1C Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:64000; Western Blot: 0.2-1 ?g/ml

Type: Primary
Antigen: CORO1C (coronin, actin binding protein, 1C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR