Anti-Coronin 1A Chicken Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7901-36EA 2410 DKK
USBIC7901-36
Anti-Coronin 1A Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:1000

Type: Primary
Antigen: Coronin 1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR