Anti-Coronavirus Surface Antigen Mouse Monoclonal Antibody [clone: 10B2297]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7901-32GEA 3910 DKK
USBIC7901-32G
Anti-Coronavirus Surface Antigen Mouse Monoclonal Antibody [clone: 10B2297]
Antistoffer
Type: Primary
Antigen: Coronavirus Surface Antigen
Clonality: Monoclonal
Clone: 10B2297
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR