Anti-Cornichon Homolog 4 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 11C275]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7901-06H-HRPEA 0 DKK
USBIC7901-06H-HRP
Anti-Cornichon Homolog 4 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 11C275]
Type: Primary
Antigen: Cornichon Homolog 4
Clonality: Monoclonal
Clone: 11C275
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR