Anti-CDSN Sheep polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7900-88EA 4780 DKK
USBIC7900-88
Anti-CDSN Sheep polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot:1 ?g/ml; Immunochemistry: 5-15 ?g/ml

Type: Primary
Antigen: CDSN (corneodesmosin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR