Anti-Core Binding Factor alpha 3 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7900-74-100EA 4650 DKK
USBIC7900-74-100
Anti-Core Binding Factor alpha 3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1-10 ?g/ml; ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: Core Binding Factor alpha 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR