Anti-Copine 6 Mouse Monoclonal Antibody [clone: 9G342]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7900-59EA 3700 DKK
USBIC7900-59
Anti-Copine 6 Mouse Monoclonal Antibody [clone: 9G342]
Antistoffer
Type: Primary
Antigen: Copine 6
Clonality: Monoclonal
Clone: 9G342
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR