Anti-Copine 1 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7900-58UEA 3460 DKK
USBIC7900-58U
Anti-Copine 1 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:16000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: Copine 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR