Anti-CPNE1 Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2588]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7900-58T-APCEA 0 DKK
USBIC7900-58T-APC
Anti-CPNE1 Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2588]
Type: Primary
Antigen: CPNE1 (copine I)
Clonality: Monoclonal
Clone: 10B2588
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR