Anti-COPS5 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7900-58JEA 3350 DKK
USBIC7900-58J
Anti-COPS5 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunofluorescence: 1:50; Western Blot: 1:1000; Immunoprecipitation: 1:50

Type: Primary
Antigen: COPS5 (COP9 signalosome subunit 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Horse, Human, Monkey, Mouse, Pig, Rat, Xenopus laevis, Zebrafish
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR