Anti-COPS5 Mouse Monoclonal Antibody [clone: 2A10.8]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7900-58CEA 4320 DKK
USBIC7900-58C
Anti-COPS5 Mouse Monoclonal Antibody [clone: 2A10.8]
Antistoffer
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: COPS5 (COP9 signalosome subunit 5)
Clonality: Monoclonal
Clone: 2A10.8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR