Anti-COP 1, alpha Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7900-45EA 4590 DKK
USBIC7900-45
Anti-COP 1, alpha Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: COP 1, alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR