Anti-CNTN6 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7865-25AEA 3700 DKK
USBIC7865-25A
Anti-CNTN6 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CNTN6 (contactin 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR