Anti-CNTN1 Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7865-19BEA 4780 DKK
USBIC7865-19B
Anti-CNTN1 Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.2?g/ml

Type: Primary
Antigen: CNTN1 (contactin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR