Anti-GJA8 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7859-06EA 2750 DKK
USBIC7859-06
Anti-GJA8 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:4000

Type: Primary
Antigen: GJA8 (gap junction protein, alpha 8, 50kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR