Anti-GJC1 Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7858-05D1BIOTEA 4620 DKK
USBIC7858-05D1BIOT
Anti-GJC1 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: GJC1 (gap junction protein, gamma 1, 45kDa)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR