Anti-GJA1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7857-02C-APEA 4620 DKK
USBIC7857-02C-AP
Anti-GJA1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: GJA1 (gap junction protein, alpha 1, 43kDa)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR