Anti-GJA1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7857-01NEA 4320 DKK
USBIC7857-01N
Anti-GJA1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 2-4?g/ml; Immunochemistry: 1:25

Type: Primary
Antigen: GJA1 (gap junction protein, alpha 1, 43kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR