Anti-GJA5 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7856-03AEA 3980 DKK
USBIC7856-03A
Anti-GJA5 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1-3?g/ml; Immunofluorescence: 1-3?g/ml; Immunochemistry: 1-3?g/ml

Type: Primary
Antigen: GJA5 (gap junction protein, alpha 5, 40kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR