Anti-GJB1 Mouse Monoclonal Antibody [clone: 10K172]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7854-05NEA 3290 DKK
USBIC7854-05N
Anti-GJB1 Mouse Monoclonal Antibody [clone: 10K172]
Antistoffer
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 1-2?g/ml

Type: Primary
Antigen: GJB1 (gap junction protein, beta 1, 32kDa)
Clonality: Monoclonal
Clone: 10K172
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR