Anti-GJB3 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2574]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7853-63-PEEA 0 DKK
USBIC7853-63-PE
Anti-GJB3 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2574]
Type: Primary
Antigen: GJB3 (gap junction protein, beta 3, 31kDa)
Clonality: Monoclonal
Clone: 10B2574
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR