Anti-Connexin 31.1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7853-61F-HRPEA 6360 DKK
USBIC7853-61F-HRP
Anti-Connexin 31.1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: Connexin 31.1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human