Anti-GJB2 Goat Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7853-04CEA 4120 DKK
USBIC7853-04C
Anti-GJB2 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:128000; Western Blot: 0. 01-0. 03?g/ml
Type: Primary
Antigen: GJB2 (gap junction protein, beta 2, 26kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat