Anti-GJB2 Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7853-04A-BIOTEA 4620 DKK
USBIC7853-04A-BIOT
Anti-GJB2 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: GJB2 (gap junction protein, beta 2, 26kDa)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR